Merck
One-step synthesis of [16]helicene.
Angewandte Chemie (International ed. in English) (2015-04-25)
Kazuyuki Mori, Takashi Murase, Makoto Fujita
ABSTRACT
A single-strand arylene-vinylene precursor containing four phenylene and three naphthylene units linked together with six vinylene spacers undergoes helical folding via sextuple photocyclization to give a [16]helicene core in a single step. The phenylene and naphthylene units are arranged in the precursor such that unfavorable side reactions (anthracene or benzoperylene formation) are avoided, and this is the key to the success of the one-step synthesis of [16]helicene, which is the longest [n]helicene that has been synthesized to date.
Sigma-Aldrich
9-Nitroanthracene, 93%