Skip to Content
Merck
All Photos(1)

Key Documents

SAAR190686

Hydrochloric acid solution

c(HCL) = 1.0mol/l (1.0 N)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
Pricing and availability is not currently available.

Product Name

Hydrochloric acid 1N solution, density: 1.02 g/mL

packaging

poly bottle of 1 × 5 L (SAAR190686LL)

density

1.02 g/mL

storage temp.

2-30°C

Application

For laboratory use only

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service