Skip to Content
Merck
All Photos(1)

Key Documents

94743

Supelco

2H,2H,3H,3H-Perfluorooctanoic acid

new

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4,4,5,5,6,6,7,7,8,8,8-Undecafluorooctanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

94743-10MG
$101.00

$101.00


Estimated to ship on17 March 2025Details



Select a Size

Change View
94743-10MG
$101.00

About This Item

Empirical Formula (Hill Notation):
C8H5F11O2
CAS Number:
Molecular Weight:
342.11
MDL number:
UNSPSC Code:
41116107

$101.00


Estimated to ship on17 March 2025Details


grade

certified reference material

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

LC/MS: suitable

application(s)

clinical testing
coatings
environmental
food and beverages

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCC(=O)O

InChI

1S/C8H5F11O2/c9-4(10,2-1-3(20)21)5(11,12)6(13,14)7(15,16)8(17,18)19/h1-2H2,(H,20,21)

InChI key

ABFCFCPCGMHSRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service