Skip to Content
Merck
All Photos(1)

Documents

749-0798

GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI-18 DSK

suitable for GC, application for SPE

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: acidified drinking water
SPE tube/cartridge: ENVI-18 DSK SPE Disk, 47 mm (57171)
condition: 5 mL dichloromethane; 5 mL methanol; 5 mL DI water
sample addition: 5 mL water sample
elution: 2 x 5 mL acetonitrile rinsing internal chamber of sample reservoir thoroughly
column: 5% phenyl polysiloxane, 30 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, SLB-5ms, 28471-U)
oven: 40 °C to 160 °C (3 min), then to 300 °C (3 min) at 6 °C/min
detector: MS, scan range m/z = 45-450
carrier gas: helium, 33 cm/sec
injection: 1 μL split/splitless, 1 min delay

suitability

application for SPE

application(s)

environmental
food and beverages

Application

Sample Pre-treatment:
Adjust 1 L drinking water to pH <2 with 6 N HCl. Add 5 mL methanol and mix thoroughly

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    2,2′,3′,4,6-Pentachlorobiphenyl
    2-Chlorobiphenyl
    Chrysene
    2,2′,3,3′,4,4′,6-Heptachlorobiphenyl
    Diethyl phthalate
    Simazine
    Hexachlorocyclopentadiene
    2,3-Dichlorobiphenyl
    Alachlor
    α-Chlordane
    Atrazine
    Fluorene
    Aldrin
    2,2′,4,4′-Tetrachlorobiphenyl
    Anthracene
    Benzo[b]fluoranthene
    Dibenz[a,h]anthracene
    Hexachlorobenzene
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    trans-Nonachlor
    Benzo[k]fluoranthene
    Pentachlorophenol
    Perylene-d12
    Acenaphthylene
    2,2′,3,3′,4,5′,6,6′-Octachlorobiphenyl
    Heptachlor
    Di-n-butyl phthalate, analytical standard, ampule of 5000 mg
    Methoxychlor
    Benzo[a]pyrene
    Dimethyl phthalate
    2,4′,5-Trichlorobiphenyl
    Heptachlor epoxide, PESTANAL®, analytical standard, isomer B
    γ-Chlordane
    Acenaphthene-d10
    Pyrene
    2,2′,4,4′,5,6′-Hexachlorobiphenyl
    Indeno[1,2,3-cd]pyrene
    Endrin
    Phenanthrene
    Benz[a]anthracene
    See All (40)

analytical column

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service