Skip to Content
Merck
All Photos(1)

Key Documents

48551

Supelco

2-Methyl-4,6-dinitrophenol

analytical standard

Synonym(s):

3,5-Dinitro-2-hydroxytoluene, 4,6-Dinitro-2-methylphenol, 4,6-Dinitro-o-cresol, DNOC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2O5
CAS Number:
Molecular Weight:
198.13
Beilstein:
2054389
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

Cc1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10H,1H3

InChI key

ZXVONLUNISGICL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-4,6-dinitrophenol may be used as an analytical reference standard for the determination of the analyte in drinking water[1] and environmental samples[2] by capillary electrophoresis and reverse-phase high-performance liquid chromatography, respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phenolic pollutants in drinking water by capillary electrophoresis in the sample stacking mode.
Rodri I, et al.
Journal of Chromatography A, 778(1-2), 279-288 (1997)
Determination of phenols in environmental samples by liquid chromatography-electrochemistry.
Cardellicchio N, et al.
Fresenius Journal of Analytical Chemistry, 358(6), 749-754 (1997)
Nicholas J Kotloski et al.
mBio, 4(1), doi:10-doi:10 (2013-01-17)
Shewanella oneidensis strain MR-1 is widely studied for its ability to respire a diverse array of soluble and insoluble electron acceptors. The ability to breathe insoluble substrates is defined as extracellular electron transfer and can occur via direct contact or
Xia Zeng et al.
Journal of agricultural and food chemistry, 57(9), 3689-3694 (2009-03-31)
The Fenton degradation of 4,6-dinitro-o-cresol (DNOC) was studied under different experimental conditions using Amberlyst 15 ion-exchange resin containing ferrous ion. DNOC was found to be effectively degraded under most conditions, and it was observed that, with the addition of HCl
Elvira Romero et al.
Biochimica et biophysica acta, 1824(6), 850-857 (2012-04-03)
Aspergillus fumigatus siderophore A (Af SidA) is a flavin-dependent monooxygenase that catalyzes the hydroxylation of ornithine, producing N(5)-hydroxyornithine. This is the first step in the biosynthesis of hydroxamate-containing siderophores in A. fumigatus. Af SidA is essential for virulence, validating this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service