Key Documents
22517
Supelco
Supelco Ferrule Nut Adapter (fits Agilent MS detectors), knurled fingertight version
pkg of 2 ea
About This Item
UNSPSC Code:
41121815
packaging
pkg of 2 ea
General description
These brass adapters enable you to use long or general purpose ferrules in Agilent MSDs. Made of brass to prevent damage to MSD source threads. Knurled head allows tightening with fingers, eliminating the need to locate the proper wrench.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Debora Keller et al.
Nanoscale, 12(44), 22511-22517 (2020-11-12)
Elementary atomic mechanisms underlying nanoparticle growth in liquids are largely unexplored and mostly a subject of conjectures based on theory and indirect experimental insights. Direct, experimental observation of such processes at an atomic level requires imaging with single-atom sensitivity and