Key Documents
Z762792
Eppendorf® Combitips advanced® pipette tips, Biopur® grade
maximum volume 5.0 mL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
blue tip
sterility
sterile
feature
barrier: no
packaging
pack of 100 ea individually wrapped
manufacturer/tradename
Eppendorf® 0030089669
maximum volume
5.0 mL
General description
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
Features and Benefits
  • pyrogen-free
  • free of human DNA
  • free of bacterial DNA
  • RNase-free
  • PCR inhibitor-free
  • DNase-free
  • ATP-free
Other Notes
Browse all pipette tips
Legal Information
Biopur is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)