Skip to Content
Merck
All Photos(1)

Key Documents

Z688878

Nunc® Skin Graft cell culture dish

four 3’ x 3’ growth chambers, 8/cs

Synonym(s):

cell culture dish, chambered cell culture dish, skin graft cell culture dish, tissue culture dish

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812

material

polystyrene

sterility

sterile

feature

lid: no

packaging

case of 8

manufacturer/tradename

Nunc 149001

dish L × W

3 mm × 76 mm

binding type

(UpCell Surface)

General description

Nunc® Skin Graft Cell Culture Dish

The Nunc skin graft growth cell culture dish is specially designed to support growth of skin and other grafts. The four chambers in one sterile (10-6 SAL) Nunc dish increase throughput and efficiency of skin graft production.

  • Ribbed 3 x 3 in. chambers separate and stabilize grafts during production, preventing them from growing into or on top of each other
  • Filling channel between chambers allows media to be added without disrupting grafts
  • Footprint permits automated initial cell seeding onto sponges, reducing labor costs and risk of contamination while increasing consistency

Application

Specially designed to support the growth of skin and other grafts; skin graft production.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service