Skip to Content
Merck
All Photos(2)

Key Documents

SML3930

Sigma-Aldrich

SDM25N

≥98% (HPLC)

Synonym(s):

(4bS,8R,8aS,14bR)-5,6,7,8,14,14b-Hexahydro-7-(2-methyl-2-propenyl)-4,8-methanobenzofuro[2,3-a]pyrido[4,3-b]carbazole-1,8a(9H)-diol, N-2-Methylallyl-noroxymorphindole, SDM 25N, SDM-25N, TU 753, TU-753, TU753

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N2O3
CAS Number:
Molecular Weight:
414.50
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent and highly selective delta-opioid receptor (DOR) antagonist.

SDM25N (TU753) is a potent and highly selective delta-opioid receptor (DOR) antagonist (δ/κ/μ Ki = 4.7 nM/3.8 μM/7.9 μM). SDM25N is also reported to inhibit Dengue virus (DENV) that restricts genomic RNA replication (EC50 = 1.9 μM using Hela DENV2 replicon cells) by targeting the viral NS4B protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franck Touret et al.
Antiviral research, 168, 109-113 (2019-05-16)
Dengue fever is the most widespread of the human arbovirus diseases, with approximately one third of the world's population at risk of infection. Dengue viruses are members of the genus Flavivirus (family Flaviviridae) and, antigenically, they separate as four closely
T Ullrich et al.
Bioorganic & medicinal chemistry letters, 11(21), 2883-2885 (2001-10-13)
In an effort to establish the importance of the N-(2-methylallyl) substituent in the noroxymorphone series, several derivatives have been synthesized, retaining that N-substituent and modifying the delta address moiety. A few compounds showed moderate binding affinity and selectivity for the
Koen W R van Cleef et al.
Antimicrobial agents and chemotherapy, 60(4), 2554-2557 (2016-02-10)
Despite the enormous disease burden associated with dengue virus infections, a licensed antiviral drug is lacking. Here, we show that the paracetamol (acetaminophen) metabolite AM404 inhibits dengue virus replication. Moreover, we find that mutations in NS4B that were previously found
Josue Jaramillo-Polanco et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 42(16), 3316-3328 (2022-03-09)
Opioid tolerance (OT) leads to dose escalation and serious side effects, including opioid-induced hyperalgesia (OIH). We sought to better understand the mechanisms underlying this event in the gastrointestinal tract. Chronic in vivo administration of morphine by intraperitoneal injection in male
Koen W R van Cleef et al.
Antiviral research, 99(2), 165-171 (2013-06-06)
Dengue virus (DENV) is an important human arthropod-borne virus with a major impact on public health. Nevertheless, a licensed vaccine or specific treatment is still lacking. We therefore screened the NIH Clinical Collection (NCC), a library of drug-like small molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service