Skip to Content
Merck
All Photos(1)

Key Documents

SML0265

Sigma-Aldrich

BIP-135

≥95% (HPLC)

Synonym(s):

3-(5-bromo-1-methyl-1H-indol-3-yl)-4-(benzofuran-3-yl)pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13BrN2O3
CAS Number:
Molecular Weight:
421.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

Cn1cc(C2=C(C(=O)NC2=O)c3coc4ccccc34)c5cc(Br)ccc15

InChI

1S/C21H13BrN2O3/c1-24-9-14(13-8-11(22)6-7-16(13)24)18-19(21(26)23-20(18)25)15-10-27-17-5-3-2-4-12(15)17/h2-10H,1H3,(H,23,25,26)

InChI key

QKQJCKAXFJBYKJ-UHFFFAOYSA-N

Biochem/physiol Actions

BIP-135 is a potent inhibitor of glycogen synthase kinase-3 (GSK3) found to be relatively selective for GSK-3β (21 nM) over GSK-3α with modest activity toward PKCβ (β1: 980 nM; β2: 219 nM), DYRK1B (590 nM), and PI3Kα (870 nM). BIP-135 was found to be a superior neuroprotective agent in a cortical neuron model of oxidative stress over other GSK-3 inhibitors, such as AR-A011418 and SB216763. In a mouse model of spinal muscular atrophy (SMA), BIP-135 elevated SMN protein levels in vitro and extended median survival.
Potent GSK3 inhibitor; neuroprotective

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service