P8648
1,2-Dinonanoyl-sn-glycero-3-phosphocholine
20 mg/mL in chloroform, ~99% (TLC)
Synonym(s):
L-α-Phosphatidylcholine, dinonanoyl
About This Item
Assay
~99% (TLC)
form
liquid
concentration
20 mg/mL in chloroform
functional group
amine
ester
storage temp.
−20°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCC)=O)COC(CCCCCCCC)=O)=O
InChI
1S/C26H53NO8P/c1-6-8-10-12-14-16-18-25(28)32-22-24(35-26(29)19-17-15-13-11-9-7-2)23-34-36(30,31)33-21-20-27(3,4)5/h24H,6-23H2,1-5H3,(H,30,31)
InChI key
HYQZGTSCHQFLRJ-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service