Skip to Content
Merck
All Photos(1)

Documents

MSST0063

Sigma-Aldrich

SILuProt IGF-1, Insulin Growth Factor -1 human

recombinant, expressed in E. coli, SIL MS Protein Standard, 15N-labeled

Synonym(s):

IGF-A, Human, Insulin Growth Factor-I, Human, Insulin-Like Growth Factor-I, Human, Insulin-like growth factor I, Myotrophin, SM-C, Somatomedin 1, Sulfation Factor C, rhIGF-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

≥97% (Heavy amino acids incorporation efficiency by MS)

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

SILuProt IGF-1 is a recombinant, stable 15N isotope-labeled human IGF-1. Expressed in E. coli, it is designed to be used as an internal standard for bioanalysis of IGF-1 in mass-spectrometry. SILuProt IGF-1 is a protein of 70 amino acids, with a calculated molecular mass of 7.74 kDa.

Biochem/physiol Actions

Insulin-like growth factor 1 (IGF-1), also called somatomedin C, is a protein that in humans is encoded by the IGF1 gene. IGF-1 is a hormone similar in molecular structure to insulin. It plays an important role in childhood growth and continues to have anabolic effects in adults. A synthetic analog of IGF-1, mecasermin, is used for the treatment of growth failure.

Sequence

GPETLCGAELVDALQFVCGDRGFYFNKPTGYGSSSRRAPQTGIVDECCFRSCDLRRLEMYCAPLKPAKSA

Physical form

Supplied as a lyophilized powder containing tris buffered saline and methionine

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service