Skip to Content
Merck
All Photos(1)

Key Documents

L6884

Sigma-Aldrich

[D-Ala6, N-Me-Leu7]-LH-RH

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H79N17O13
CAS Number:
Molecular Weight:
1210.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(N(C)C(=O)C(C)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CO)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(Cc4cnc[nH]4)NC(=O)C5CCC(=O)N5)C(=O)NC(CCCNC(N)=N)C(=O)N6CCCC6C(=O)NCC(N)=O

Amino Acid Sequence

pGlu-His-Trp-Ser-Tyr-D-Ala-N-Methyl-Leu-Arg-Pro-Gly-NH2

Biochem/physiol Actions

An LH-RH analog with over 5× the potency of LH-RH

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analogs of luteinizing hormone releasing factor (LRF). Synthesis and biological activity of ((Nalpha-Me)Leu-7)LRF and (D-Ala-6,(Nalpha-Me)Leu-7)LRF.
N Ling et al.
Biochemical and biophysical research communications, 63(3), 801-806 (1975-04-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service