Skip to Content
Merck
All Photos(1)

Key Documents

L5257

Sigma-Aldrich

1-Heptadecanoyl-sn-glycero-3-phosphocholine

~99%

Synonym(s):

L-α-Lysophosphatidylcholine, heptadecanoyl, Lysolecithin, heptadecanoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H52NO7P
CAS Number:
Molecular Weight:
509.66
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)COP([O-])(=O)OCC[N+](C)(C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Jeffrey North et al.
Bioorganic & medicinal chemistry, 17(9), 3433-3442 (2009-04-07)
Autotaxin (ATX) catalyzes the hydrolysis of lysophosphatidylcholine (LPC) to form the bioactive lipid lysophosphatidic acid (LPA). LPA stimulates cell proliferation, cell survival, and cell migration and is involved in obesity, rheumatoid arthritis, neuropathic pain, atherosclerosis and various cancers, suggesting that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service