Skip to Content
Merck
All Photos(1)

Key Documents

L1289

Sigma-Aldrich

Lavendustin C

≥97%, solid

Synonym(s):

N-(2′,5′-Dihydroxybenzyl)-5-aminosalicylic acid, Compound 5, HDBA, [2-Hydroxy-5-[N-(2′,5′-dihydroxybenzyl)amino]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO5
CAS Number:
Molecular Weight:
275.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97%

form

solid

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

OC(=O)c1cc(NCc2cc(O)ccc2O)ccc1O

InChI

1S/C14H13NO5/c16-10-2-4-12(17)8(5-10)7-15-9-1-3-13(18)11(6-9)14(19)20/h1-6,15-18H,7H2,(H,19,20)

InChI key

LULATDWLDJOKCX-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Potent inhibitor of protein tyrosine kinases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desireé Alesa Gyles et al.
Polymers, 12(3) (2020-03-25)
Chronic wounds are severe breaks in the skin barrier that fail to heal in an acceptable time-frame, thus preventing the complete restoration of the tissue's anatomical and functional integrity, increasing the likelihood of infections and apoptosis. Hydrogels are known as
Wenwen Zhang et al.
International journal of medical sciences, 10(13), 1888-1898 (2013-12-11)
Aberrant activation of β-catenin signaling plays an important role in human tumorigenesis. However, molecular mechanisms behind the β-catenin signaling deregulation are mostly unknown because genetic alterations in this pathway only account for a small fraction of tumors. Here, we investigator
Liz Valle-Aviles et al.
BMC microbiology, 7, 107-107 (2007-12-01)
Sporothrix schenckii is a pathogenic, dimorphic fungus, the etiological agent of sporotrichosis, a subcutaneous lymphatic mycosis. Dimorphism in S. schenckii responds to second messengers such as cAMP and calcium, suggesting the possible involvement of a calcium/calmodulin kinase in its regulation.
J J H Chu et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(9), 3520-3525 (2007-03-16)
Dengue virus is a mosquito-borne flavivirus that represents an important emerging infectious disease and is an international health concern. Currently, there is no vaccine or effective antiviral therapy to prevent or to treat dengue virus infection. The slow progress in
Jieun Yoon et al.
Bioorganic & medicinal chemistry letters, 20(13), 3930-3935 (2010-06-24)
Lavendustin-mimetic small molecules modifying the linker -CH(2)-NH- with an 1,2,3-triazole ring have been synthesized via a click chemistry. Two pharmacophoric fragments of lavendustin were varied to investigate chemical space and the auxophoric -CH(2)-NH- was altered to an 1,2,3-triazole for rapid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service