Skip to Content
Merck
All Photos(1)

Key Documents

I1661

Sigma-Aldrich

ICA-105574

≥98% (HPLC)

Synonym(s):

3-nitro-N-(4-phenoxyphenyl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N2O4
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

[O-][N+](=O)c1cccc(c1)C(=O)Nc2ccc(Oc3ccccc3)cc2

InChI

1S/C19H14N2O4/c22-19(14-5-4-6-16(13-14)21(23)24)20-15-9-11-18(12-10-15)25-17-7-2-1-3-8-17/h1-13H,(H,20,22)

InChI key

GDWKBKTVROCPNZ-UHFFFAOYSA-N

Biochem/physiol Actions

ICA-105574 is a potent and efficacious hERG potassium channel activator, that removes hERG inactivation.
hERG (Kv11.1) is a voltage-gated potassium channel with important cardiovascular function. Many potential and marketed drugs bind to and inhibit hERG function, causing prolongation of the electrocardiogram QT interval, which leads to life-threatening ventricular arrhythmias. Understanding of hERG function is critical for drug discovery and development. hERG exhibits a unique bell-shaped current-voltage relationship which is a result of very rapid inactivation of the channel upon voltage activation. ICA-105574 is a small molecule activator of hERG that binds to the channel to remove inactivation, thereby increasing peak current amplitude and shortening the action potential. It also modulates activation kinetics of the channel. ICA-105574 differs in efficacy, mechanism of action, and/or binding site from other known hERG activators and enhancers. This compound is a valuable tool for furthering understanding of hERG biophysics and physiology, expecially the structural transitions that occur during inactivation.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jared N Tschirhart et al.
Molecular pharmacology, 95(4), 386-397 (2019-01-23)
The human ether-a-go-go-related gene (hERG) encodes the pore-forming subunit of the rapidly activating delayed rectifier potassium channel (IKr). Drug-mediated or medical condition-mediated disruption of hERG function is the primary cause of acquired long-QT syndrome, which predisposes affected individuals to ventricular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service