Skip to Content
Merck
All Photos(1)

Key Documents

I0762

Sigma-Aldrich

(S)-(+)-1-Iodo-2-methylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH2I
CAS Number:
Molecular Weight:
198.05
MDL number:
UNSPSC Code:
12352000
Pricing and availability is not currently available.

contains

copper as stabilizer

refractive index

n20/D 1.497 (lit.)

bp

148 °C (lit.)

solubility

ethanol: soluble, clear to slightly hazy

density

1.525 g/mL at 25 °C (lit.)

SMILES string

CC[C@H](C)CI

InChI

1S/C5H11I/c1-3-5(2)4-6/h5H,3-4H2,1-2H3/t5-/m0/s1

InChI key

RHBHXHXNWHTGSO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service