Skip to Content
Merck
All Photos(6)

Documents

HPA040503

Sigma-Aldrich

Anti-CEP192 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Centrosomal protein 192kDa, Anti-FLJ10352, Anti-KIAA1569

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FVLKERTQENVTLIYNPSDRGINNKTATELSTVYLFGGDEISRQQYRRALLHKPEMIKQILPEHSVLQNINFVEAFQDELLVTEVYD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEP192(55125)

General description

Centrosomal protein 192 (CEP192) is a 192 kDa protein present in the centrosome. Cep192 is located on human chromosome 18p11.21.

Immunogen

centrosomal protein 192kDa recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Centrosomal protein 192 (CEP192) plays key role in the formation of spindle fibres during mitosis. CEP192 coordinates with CEP152 for the recruitment of polo-like kinase 4 (Plk4) on centrioles during cell cycle. CEP192 also interacts with Aurora A kinase and modulates the centrosome maturation and spindle assembly pathway. Mutation the CEP192 at proline residue leads to impairment in functional spindle fibre formation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81634

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Cep192-organized aurora A-Plk1 cascade is essential for centrosome cycle and bipolar spindle assembly
Joukov V, et al.
Molecular Cell, 55(4), 578-591 (2014)
Human Cep192 and Cep152 cooperate in Plk4 recruitment and centriole duplication
Sonnen KF, et al.
Journal of Cell Science, jcs-129502 (2013)
Cep192 controls the balance of centrosome and non-centrosomal microtubules during interphase
O?Rourke BP, et al.
PLoS ONE, 9(6), e101001-e101001 (2014)
GWAS of 972 autologous stem cell recipients with multiple myeloma identifies 11 genetic variants associated with chemotherapy-induced oral mucositis
Coleman EA, et al.
Supportive Care in Cancer : Official Journal of the Multinational Association of Supportive Care in Cancer, 23(3), 841-849 (2015)
PHD1 links cell-cycle progression to oxygen sensing through hydroxylation of the centrosomal protein Cep192
Moser SC, et al.
Developmental Cell, 26(4), 381-392 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service