recombinant fusion protein corresponding to residues 119-155 of human HCN4 fused to GST.
Physical form
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Microbiology and immunology, 66(7), 353-360 (2022-05-08)
The adverse factors impacting the intestinal microbiota of newborns remain to be elucidated. We put forward a hypothesis that hyperoxia in combination with rituximab exhibits a synergistic effect that interferes with neonatal intestinal microbiota. Six C57BL/6J mice, aged 12 weeks
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.