Skip to Content
Merck
All Photos(1)

Key Documents

EP022492209

Eppendorf® epT.I.P.S.® Sterile, singles

volume 2-200 μL, L 53 mm, yellow, colorless tip, pkg of 1000 ea (individually packed)

Synonym(s):

pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600

material

colorless tip
polypropylene
yellow

sterility

sterile

feature

endotoxin free

packaging

pkg of 1000 ea (individually packed)

manufacturer/tradename

Eppendorf® 022492209

L

53 mm

volume

2-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

epT.I.P.S.® Singles, sterile, 2 – 200 µL, 53 mm, yellow, colorless tips, 1,000 tips, individually packed
  • For use in medical, pharmaceutical and food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by independent laboratory
  • Lot-specific certificates available at www.eppendorf.com/certificates

Features and Benefits

  • For use in medical, pharmaceutical and food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by independent laboratory
  • Lot-specific certificates available at www.eppendorf.com/certificates

  • For use in medical, pharmaceutical and food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by independent laboratory
  • Lot-specific certificates available at www.eppendorf.com/certificates

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service