D0413
1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine
~97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
~97%
refractive index
n20/D 1.565 (lit.)
bp
95 °C/0.7 mmHg (lit.)
density
1.054 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CN1CCN(C)P1c2ccccc2
Looking for similar products? Visit Product Comparison Guide
Application
Reagent for the stereospecific synthesis of olefins from 1,2-diols.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
221.0 °F - closed cup
Flash Point(C)
105 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron Letters, 23, 1979-1979 (1982)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service