Documents
BAH378620102
SP Bel-Art®Black Pen
Black Felt Tip Pen
About This Item
UNSPSC Code:
41104000
NACRES:
NB.22
manufacturer/tradename
Bel-Art H37862-0102
General description
A cost effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate.
Features and Benefits
  • Fast, accurate and economical sample counting and sample count verification
  • Marks, counts, and gives an audible and visual alert each time a count is registered
  • Lightweight, pen-style design reduces the likelihood of error; combines all aspects of counting into one motion
  • Stores up to 35 sample counts
  • Verify sample counts with up/down counting
  • Displays sum total of all colonies counted
  • Backlit display for easy viewing in low light conditions
  • Audible alert can be turned off if desired
  • Counting function can be locked to prevent error during breaks between counting
  • Long life, red and black felt-tip markers (included) work on plastic or glass and are oil and water resistant; ink is removable with ethyl alcohol
  • Includes 3V lithium battery and protective storage case
  • For a complete Colony Counter System, see catalog no. F37863-3000
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP