Skip to Content
Merck
All Photos(1)

Key Documents

B3882

Sigma-Aldrich

Arachidyl behenate

Synonym(s):

Behenic acid arachidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84O2
CAS Number:
Molecular Weight:
621.12
EC Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCCCCCC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service