Skip to Content
Merck
All Photos(1)

Key Documents

AV48994

Sigma-Aldrich

Anti-CHST14 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human, dog, rat, mouse, pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CHST14(113189)

Immunogen

Synthetic peptide directed towards the middle region of human CHST14

Application

Anti-CHST14 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Carbohydrate (N-acetylgalactosamine 4-0-sulfotransferase 14, CHST14; D4ST1) catalyzes the transfer of sulfate to the C-4 hydroxyl of N-acetylgalactosamine residues in dermatan sulfate.CHST14 regulates the proliferation and neurogenesis process in neural progenitor cells.[1] Mutations in CHST14 gene causes adducted thumb-clubfoot syndrome.[2]

Sequence

Synthetic peptide located within the following region: REYQQRYGAEIVRRYRAGAGPSPAGDDVTFPEFLRYLVDEDPERMNEHWM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Munis Dündar et al.
American journal of human genetics, 85(6), 873-882 (2009-12-17)
Adducted thumb-clubfoot syndrome is an autosomal-recessive disorder characterized by typical facial appearance, wasted build, thin and translucent skin, congenital contractures of thumbs and feet, joint instability, facial clefting, and coagulopathy, as well as heart, kidney, or intestinal defects. We elucidated
Shan Bian et al.
Journal of cell science, 124(Pt 23), 4051-4063 (2011-12-14)
Chondroitin sulfates (CSs) and dermatan sulfates (DSs) are enriched in the microenvironment of neural stem cells (NSCs) during development and in the adult neurogenic niche, and have been implicated in mechanisms governing neural precursor migration, proliferation and differentiation. In contrast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service