A2473
ART® self-sealing barrier pipette tips
ART 10 Reach, volume 0.1-10 μL, sterile
All Photos(2)
About This Item
UNSPSC Code:
41100000
sterility
sterile
packaging
case of 5 packs
pkg of 960 tips (racked)
volume measuring range
0.1-10 μL
General description
Extended tip length of the Art 10 Reach prevents contamination of samples, tubes and pipetters.
Packaging
960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Compatibility
Compatibility
Biohit mLINE .5-10 μl
Biohit Electronic .2-10 μl
Biohit Variable Single/Multi-Channel .5-10 μl
Capp Aero 10 μl
Corning 4959 .1-2 μl
Corning 4960 .5-10 μl
Eppendorf Reference/Research .1-2.5 μl
Finnpipette 4500 Digital .2-2 μl
Finnpipette 4500 Digital Micro .5-10 μl
Finnpipette Focus 4600 3ul, 5ul, 10 μl
Finnpipette Multi-Channel .5-10 μl
Hamilton SoftGrip Precision wul/10 μl
Mettler-Toledo VoluMate .1-2.5 μl
Nichiryo BenchMate .2-2ul/.5-10 μl
Pipetman P-2 .1-2 μl
Pipetman P-10 .5-10 μl
Rainin EDP 2 .5-10 μl
Rainin EDP Plus .5-10 μl
Rainin Pipet Plus .2-2ul/.5-10 μl
Rainin Pipetman .2-2ul/.5-10 μl
Transferpette Single/Multi-Channel .5-10ul/2-20 μl
Transferpette Ultra-Micro 2-20 μl
Biohit mLINE .5-10 μl
Biohit Electronic .2-10 μl
Biohit Variable Single/Multi-Channel .5-10 μl
Capp Aero 10 μl
Corning 4959 .1-2 μl
Corning 4960 .5-10 μl
Eppendorf Reference/Research .1-2.5 μl
Finnpipette 4500 Digital .2-2 μl
Finnpipette 4500 Digital Micro .5-10 μl
Finnpipette Focus 4600 3ul, 5ul, 10 μl
Finnpipette Multi-Channel .5-10 μl
Hamilton SoftGrip Precision wul/10 μl
Mettler-Toledo VoluMate .1-2.5 μl
Nichiryo BenchMate .2-2ul/.5-10 μl
Pipetman P-2 .1-2 μl
Pipetman P-10 .5-10 μl
Rainin EDP 2 .5-10 μl
Rainin EDP Plus .5-10 μl
Rainin Pipet Plus .2-2ul/.5-10 μl
Rainin Pipetman .2-2ul/.5-10 μl
Transferpette Single/Multi-Channel .5-10ul/2-20 μl
Transferpette Ultra-Micro 2-20 μl
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)