Skip to Content
Merck
All Photos(1)

Key Documents

54298

Sigma-Aldrich

(−)-Nortrachelogenin

≥95.0% (HPLC)

Synonym(s):

(−)-Wikstromol, (3S)-4,5-Dihydro-4β-(4-hydroxy-3-methoxybenzyl)-3-(4-hydroxy-3-methoxybenzyl)-3β-hydroxyfuran-2(3H)-one, (3S,4S)-3-Hydroxy-3,4-bis(4-hydroxy-3-methoxybenzyl)dihydrofuran-2-one, (3S,4S)-Dihydro-3-hydroxy-3,4-bis[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, NSC 271296, Pinopalustrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O7
CAS Number:
Molecular Weight:
374.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (HPLC)

SMILES string

COc1cc(C[C@H]2COC(=O)[C@]2(O)Cc3ccc(O)c(OC)c3)ccc1O

InChI

1S/C20H22O7/c1-25-17-8-12(3-5-15(17)21)7-14-11-27-19(23)20(14,24)10-13-4-6-16(22)18(9-13)26-2/h3-6,8-9,14,21-22,24H,7,10-11H2,1-2H3/t14-,20-/m0/s1

InChI key

ZITBJWXLODLDRH-XOBRGWDASA-N

Application

Nortrachelogenin [(−)-Wikstromol], a bioactive α-hydroxylated lactone, is a pharmacologically active lignan used to study its antitumor activities versus breast and lung cancer cells. Nortrachelogenin may be used as a reference compound during the isolation and identification of α-hydroxylated lactone type lignans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-sheng Feng et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 38(12), 927-930 (2004-03-26)
To study the chemical constituents from the water-extracts of pine needles of Pinus massoniana Lamb. Chromatographic techniques were used to separate and purify compounds. Their physico-chemical properties and spectral data (UV, IR, MS, 1H-1H, 13C-1H NMR, DEPT, HMBC etc.) were
B S Sachin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 862(1-2), 237-241 (2008-01-15)
A specific and sensitive high-performance liquid chromatographic (HPLC) method with photodiode-array (PDA) ultraviolet detection was developed for the simultaneous determination of three bioactive constituents of Cedrus deodara namely wikstromol, matairesinol and dibenzylbutyrolactol in mouse plasma. In solid-phase extraction (SPE) these
N M Saarinen et al.
The Journal of steroid biochemistry and molecular biology, 93(2-5), 209-219 (2005-04-30)
Low risk of breast cancer (BC) has been proposed to be associated with high intake of lignans. Some plant lignans are converted to mammalian lignans, e.g., enterolactone (ENL), suggested to be the biologically active lignan forms. Until now, little attention
Emilia Peuhu et al.
Biochemical pharmacology, 86(5), 571-583 (2013-06-12)
Prostate cancer cells frequently develop resistance toward androgen-deprivation and chemotherapy. To identify new approaches to treat androgen-dependent prostate cancer, we have performed a structure-activity analysis of lignan polyphenols for cancer cell specific sensitization to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)
Xing-Qi Tan et al.
Planta medica, 71(1), 93-95 (2005-01-29)
Seven lignans having a diarylhydroxybutyrolactone skeleton were isolated from the leaves and stems of Trachelospermum jasminoides (Lindl.) Lem. Their structures were elucidated to be nortrachelogenin 8' -O-beta-D-glucopyranoside (1), nortrachelogenin 5'- C- beta-D-glucopyranoside (2), trachelogenin amide (3), nortracheloside, trachelogenin, tracheloside, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service