Skip to Content
Merck
All Photos(1)

Key Documents

297305

Sigma-Aldrich

Formic acid-14C sodium salt

≥98% radiochemical purity basis (HPLC), aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H14COONa
CAS Number:
Molecular Weight:
68.01
MDL number:
UNSPSC Code:
12352200
Product 297305 is not currently sold in your country. Contact Technical Service

Assay

≥98% radiochemical purity basis (HPLC)

form

aqueous solution

extent of labeling

1-20 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

[H][14C](=O)O[Na]

Looking for similar products? Visit Product Comparison Guide

Packaging

Combi-vial

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service