Skip to Content
Merck
All Photos(1)

Key Documents

I5397

Supelco

Lithium ionophore VI

suitable for function test

Synonym(s):

6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane, 6,6-Dibenzyl-14-crown-4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Beilstein:
5991129
MDL number:
UNSPSC Code:
26111700

suitability

suitable for function test

storage temp.

2-8°C

SMILES string

C1COCCOCC(COCCOC1)(Cc2ccccc2)Cc3ccccc3

InChI

1S/C24H32O4/c1-3-8-22(9-4-1)18-24(19-23-10-5-2-6-11-23)20-27-16-14-25-12-7-13-26-15-17-28-21-24/h1-6,8-11H,7,12-21H2

InChI key

KDMUFHBMXZLDLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neutral carrier for Li+-ions in PVC ion-selective electrodes with excellent selectivities over potassium and sodium ions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service