95995
Z-Asp(OBzl)-OBzl
purum, ≥98.0% (TLC)
Synonym(s):
Z-L-aspartic acid dibenzyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H25NO6
CAS Number:
Molecular Weight:
447.48
Beilstein:
2067438
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
grade
purum
Assay
≥98.0% (TLC)
SMILES string
O=C(C[C@H](NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2)OCc3ccccc3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service