Skip to Content
Merck
All Photos(1)

Key Documents

93346

Sigma-Aldrich

Trizma® hemisulfate

≥99.0% (T)

Synonym(s):

Bis(tris[hydroxymethyl]aminomethane) sulfate, TRIS hemisulfate salt, Tris(hydroxymethyl)aminomethane hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · 0.5 H2SO4
CAS Number:
Molecular Weight:
170.17
Beilstein:
3718745
EC Number:
MDL number:
UNSPSC Code:
12161700

Assay

≥99.0% (T)

impurities

≤0.2% water

mp

163-166 °C

solubility

H2O: 0.5 M at 20 °C, clear

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OS(O)(=O)=O.NC(CO)(CO)CO.NC(CO)(CO)CO

InChI

1S/2C4H11NO3.H2O4S/c2*5-4(1-6,2-7)3-8;1-5(2,3)4/h2*6-8H,1-3,5H2;(H2,1,2,3,4)

InChI key

OLJBMOUMSAPNML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service