65325
8-Methoxypyrene-1,3,6-trisulfonic acid trisodium salt
for fluorescence, ≥98.0% (TLC)
Synonym(s):
PTS1, Trisodium 8-methoxypyrene-1,3,6-trisulfonate
About This Item
Recommended Products
grade
for fluorescence
Quality Level
Assay
≥98.0% (TLC)
form
solid
mp
253-260 °C (dec.) (lit.)
solubility
DMF: soluble
DMSO: soluble
H2O: soluble
methanol: soluble
fluorescence
λex 404 nm; λem 431 nm in H2O
SMILES string
[Na+].[Na+].[Na+].COc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C17H12O10S3.3Na/c1-27-12-6-13(28(18,19)20)9-4-5-11-15(30(24,25)26)7-14(29(21,22)23)10-3-2-8(12)16(9)17(10)11;;;/h2-7H,1H3,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3
InChI key
GUGNSJAORJLKGP-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service