Skip to Content
Merck
All Photos(1)

Key Documents

55122

Supelco

Propyl valerate

puriss. p.a., standard for GC, ≥99.7% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)2CH2COO(CH2)2CH3
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747358
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
Pricing and availability is not currently available.

grade

standard for GC
puriss. p.a.

Assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.407

density

0.870 g/mL at 20 °C (lit.)

application(s)

environmental

SMILES string

CCCCC(=O)OCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service