Skip to Content
Merck
All Photos(1)

Key Documents

47764

Sigma-Aldrich

(Formyloxy)acetonitrile

purum, ≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOCH2CN
CAS Number:
Molecular Weight:
85.06
Beilstein:
1741714
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (T)

refractive index

n20/D 1.407 (lit.)
n20/D 1.407

bp

173 °C (lit.)

density

1.187 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)OCC#N

InChI

1S/C3H3NO2/c4-1-2-6-3-5/h3H,2H2

InChI key

ZFLAEHBSVFWEHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the chemoselective formylation of amines and alcohols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Deutsch et al.
Synthetic Communications, 23, 1561-1561 (1993)
W. Duczek et al.
Synthesis, 37-37 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service