43117
Ethoxy-methyl-diphenylsilane
purum, ≥97.0% (GC)
Synonym(s):
Diphenyl-ethoxy-methylsilane, Diphenyl-methyl-ethoxysilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H18OSi
CAS Number:
Molecular Weight:
242.39
Beilstein:
2940692
EC Number:
MDL number:
UNSPSC Code:
12352103
grade
purum
Assay
≥97.0% (GC)
refractive index
n20/D 1.544
bp
100-102 °C/0.3 mmHg (lit.)
SMILES string
CCO[Si](C)(c1ccccc1)c2ccccc2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service