Skip to Content
Merck
All Photos(1)

Key Documents

38818

Sigma-Aldrich

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

purum, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70
Beilstein:
3634126
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (AT)

mp

260-265 °C (lit.)
260-265 °C

SMILES string

Cl.COc1cc2CCNCc2cc1OC

InChI

1S/C11H15NO2.ClH/c1-13-10-5-8-3-4-12-7-9(8)6-11(10)14-2;/h5-6,12H,3-4,7H2,1-2H3;1H

InChI key

SHOWAGCIRTUYNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for the synthesis of tetrahydroisoquinolines[1][2][3]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.I. Meyers et al.
Angewandte Chemie (International Edition in English), 96, 448-448 (1984)
Organic Syntheses, 67, 60-60 (1989)
The Journal of Organic Chemistry, 55, 4218-4218 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service