Skip to Content
Merck
All Photos(1)

Key Documents

34509

Supelco

Monuron-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

N′-(4-Chlorophenyl)-N,N-di(methyl-d3)urea

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$917.00

$917.00


Estimated to ship on31 January 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$917.00

About This Item

Empirical Formula (Hill Notation):
C9D6H5ClN2O
CAS Number:
Molecular Weight:
204.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$917.00


Estimated to ship on31 January 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=CC=C(NC(N(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C1

InChI

1S/C9H11ClN2O/c1-12(2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)/i1D3,2D3

InChI key

BMLIZLVNXIYGCK-WFGJKAKNSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betty Chaumet et al.
Ecotoxicology (London, England), 29(2), 185-195 (2020-01-24)
Variations of temperature and photoperiod throughout different seasons can affect aquatic communities such as biofilms. Biofilms, generally present at the base of trophic chains in freshwaters, are also subject to organic contamination, and are especially affected by herbicides. Many studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service