Skip to Content
Merck
All Photos(1)

Key Documents

30722

Supelco

Chromoionophore XIII

Selectophore, ≥95.0% (CHN)

Synonym(s):

7-[2-(Decyloxycarbonyl)-phenyl]-10-diethylamino-3-hydroxy-benzo[c]xanthylium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H44ClNO8
CAS Number:
Molecular Weight:
678.21
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Assay

≥95.0% (CHN)

SMILES string

[O-]Cl(=O)(=O)=O.CCCCCCCCCCOC(=O)c1ccccc1-c2c3ccc(cc3[o+]c4c2ccc5cc(O)ccc45)N(CC)CC

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Lipophilic pH indicator used to assay biogenic amines with optodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. He et al.
Analytica Chimica Acta, 246, 251-251 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service