Skip to Content
Merck
All Photos(1)

Key Documents

15130

Sigma-Aldrich

Boc-Leu-ONp

purum, ≥96.0% (TLC)

Synonym(s):

Boc-L-leucine 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N2O6
CAS Number:
Molecular Weight:
352.38
Beilstein:
1894893
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

purum

Assay

≥96.0% (TLC)

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)OC(C)(C)C)C(=O)Oc1ccc(cc1)[N+]([O-])=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cho-Hao Lin et al.
Vascular pharmacology, 63(2), 88-96 (2014-12-04)
A feature of vascular smooth muscle cells is their unique ability to exist in multiple phenotypes permitting a broad range of functions that include contraction, proliferation, or synthesis and secretion of extracellular matrix. Although it is known that these phenotypes
Timothy R Fennell et al.
Chemico-biological interactions, 227, 112-119 (2015-01-07)
2,3-Butanedione (BD) is a reactive diketone in artificial butter flavors that is thought to cause bronchiolitis obliterans in workers in microwave popcorn manufacturing. Bronchiolitis obliterans is generally not diagnosed until irreversible damage has occurred; therefore a biomarker of early exposure
Doreen Matthies et al.
Nature communications, 5, 5286-5286 (2014-11-11)
All rotary ATPases catalyse the interconversion of ATP and ADP-Pi through a mechanism that is coupled to the transmembrane flow of H(+) or Na(+). Physiologically, however, F/A-type enzymes specialize in ATP synthesis driven by downhill ion diffusion, while eukaryotic V-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service