Skip to Content
Merck
All Photos(1)

Documents

03920590

Oleanolic acid

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

mp

>300 °C (lit.)

application(s)

food and beverages

SMILES string

[H][C@@]12CC(C)(C)CC[C@@]1(CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C)C(O)=O

InChI

1S/C30H48O3/c1-25(2)14-16-30(24(32)33)17-15-28(6)19(20(30)18-25)8-9-22-27(5)12-11-23(31)26(3,4)21(27)10-13-29(22,28)7/h8,20-23,31H,9-18H2,1-7H3,(H,32,33)/t20-,21-,22+,23-,27-,28+,29+,30-/m0/s1

InChI key

MIJYXULNPSFWEK-GTOFXWBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Triterpenoid isolated from medicinal plants. Antitumor and hepatoprotective agent. Oleanolic acid has therapeutic potential for neurodegenerative diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Liangfeng Fu et al.
Organic letters, 15(7), 1622-1625 (2013-03-28)
Bardoxolone methyl (2-cyano-3,12-dioxooleane-1,9(11)-dien-28-oic acid methyl ester; CDDO-Me) (1), a synthetic oleanane triterpenoid with highly potent anti-inflammatory activity (levels below 1 nM), has completed a successful phase I clinical trial for the treatment of cancer and a successful phase II trial
Dick de Zeeuw et al.
The New England journal of medicine, 369(26), 2492-2503 (2013-11-12)
Although inhibitors of the renin-angiotensin-aldosterone system can slow the progression of diabetic kidney disease, the residual risk is high. Whether nuclear 1 factor (erythroid-derived 2)-related factor 2 activators further reduce this risk is unknown. We randomly assigned 2185 patients with
Philippe Charlier et al.
Scientific reports, 3, 1296-1296 (2013-03-02)
During the Middle Ages, the partition of the cadaver of the elite members was a current practice, with highly technical treatment given to symbolic organs such as the heart. Considered mostly from a theoretical point of view, this notion of
Min Song et al.
Journal of pharmaceutical and biomedical analysis, 40(1), 190-196 (2005-08-30)
A highly selective and sensitive HPLC-ESI-MS-MS method was developed for the determination of oleanolic acid in human plasma. The oleanolic acid and glycyrrhetinic acid (internal standard) were recovered from plasma with ethyl acetate liquid-liquid extraction. The organic extracts were dried
Fei Yu et al.
Journal of medicinal chemistry, 56(11), 4300-4319 (2013-05-15)
Development of hepatitis C virus (HCV) entry inhibitors represents an emerging approach that satisfies a tandem mechanism for use with other inhibitors in a multifaceted cocktail. By screening Chinese herbal extracts, oleanolic acid (OA) was found to display weak potency

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service