Skip to Content
Merck
All Photos(1)

Documents

MXRPCNA1K2K

Millipore

Power Cord, Power MIX 1000/2000 Control Box, NA, 125V, 20A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103801
eCl@ss:
32010600

AC/DC input

125 V, 20 A

Quality Level

compatibility

for use with MOBIUS® Power MIX 1000 and 2000

Related Categories

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service