Skip to Content
Merck
All Photos(1)

Documents

MABS1962

Sigma-Aldrich

Anti-PEG Antibody, clone rAGP6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

antibody form

purified antibody

clone

monoclonal
rAGP6

purified by

(Purified by size-exclusion chromatography)

species reactivity

all, all, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable

isotype

IgMκ

General description

PEG-conjugated to E.coli beta-glucuronidase.

Specificity

Clone rAGP6 is a rat IgM monoclonal antibody with specificity for the repeating subunits present in PEG. rAGP6 displays high affinity to both short and long PEG molecules.

Quality

Evaluated by ELISA with PEG molecules (10K-PEG, 3K-PEG, and 2K-mPEG).

ELISA Analysis: 5 µg/mL of Polyethylene Glycol (PEG) (10K-PEG, 3K-PEG, and 2K-mPEG)were detected by this antibody.

Physical form

Purified rat monoclonal antibody IgM in PBS with 0.04% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service