Skip to Content
Merck
All Photos(2)

Documents

MAB8151-I

Sigma-Aldrich

Anti-West Nile Virus/Kunjin Antibody, Envelope Antibody, clone 3.91D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3.91D, monoclonal

mol wt

calculated mol wt 31.89 kDa
observed mol wt ~50 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG3

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

vaccinia virus ... poly> POLY(912267)

Related Categories

General description

Genome polyprotein (UniProt: P14335) is encoded by the POLY gene (Gene ID: 912267) in West Nile virus. West Nile virus (WNV) is a mosquito-borne positive-sense RNA virus of the Flavivirus genus within the Flaviviridae family. WNV genome encodes for three structural domains known as C, prM, and E and seven non-structural proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B, and NS5). These are translated as a single polypeptide, which is subsequently cleaved by viral and cellular proteases. The envelope glycoprotein (E) undergoes a conformational rearrangement triggered by low pH that results in a class II fusion event required for viral entry. The E protein (aa 291-791) contains a three-domain structure where domain I is an eight-stranded β-barrel in the center portion of the protein (aa 1-51, 134-195, and 284-297). The DII domain consists of 170 residues inserted into DI (aa 52-133 and 196-283). At the distal end of DII domain is a fusion loop (aa 98-110) and a conserved glycine-rich, hydrophobic sequence, which is important for flaviviral fusion. The third domain, DIII, is an Ig-like domain with seven β-strand and has been implicated in receptor binding. The E protein also has a single N-linked glycosylation site at Asn 154, located within the ectodomain of prM domain and is cleaved by cellular enzymes during viral particle maturation. Clone E60 specifically binds to the DII fusion loop and displays limited neutralizing activity and even at higher doses neutralization activity is shown to be between 26 and 68%. (Ref.: Martín-Acebes MA, et al. (2012). World J Virol. 1(2); 51-70).

Specificity

Clone 3.91D is a mouse monoclonal antibody that detects West Nile virus envelope protein. It targets an epitope within the N-terminal half of polyprotein.

Immunogen

West Nile Virus (WNV; subtype KUNV) isolate OR393.

Application

Qualtiy Control Testing

Evaluated by Western Blotting with recombinant West Nile Virus envelope protein.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected recombinant West Nile Virus envelope protein.

Tested Applications


Western Blotting Analysis: A representative lot detected West Nile Virus/Kunjin, Envelope protein in Western Blotting applications (Maeda, A., et al. (2009). Virus Res.;144(1-2):35-43; Saiyasombat, R., et al. (2014). Virol J.;11:150; Blitvich, B.J., et al. (2016). Am J Trop Med Hyg.;95(5):1185-1191).

ELISA Analysis: Various dilutions of this antibody detected recombinant West Nile Virus/Kunjin, Envelope protein.

Immunofluorescence Analysis: A representative lot detected West Nile Virus/Kunjin, Envelope protein in Immunofluorescence applications (Maeda, A., et al. (2009). Virus Res.;144(1-2):35-43; Osorio, J.E., et al. (2012). Am J Trop Med Hyg.;87(3):565-72).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-West Nile Virus/Kunjin, Envelope, clone 3.91D, Cat. No. MAB8151-I, is a mouse monoclonal antibody that detects envelope protein in West Nile virus and is tested for use in ELISA, Immunofluorescence, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service