Skip to Content
Merck
All Photos(1)

Key Documents

MAB325

Sigma-Aldrich

Anti-Syntaxin Antibody, clone SP7

purified antibody, clone SP7, Chemicon®

Synonym(s):

Syntaxin 1A, Neuron-specific Antigen HPC-1, STX1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

SP7, monoclonal

species reactivity

hamster, pig, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STX1A(6804)

Specificity

Syntaxin (34-36 kDa). Reacts with syntaxin expressed in bacterial systems. Useful to investigators who transplant human neurons or human neural derived cell lines into rodents since the antibody reacts with human but poorly with rat syntaxin. Also useful in double labeling studies due to the fact it is an IgG2a subclass monoclonal.

Immunogen

Crude synaptic immunoprecipitate (human).

Application

Anti-Syntaxin Antibody, clone SP7 is an antibody against Syntaxin for use in ELISA, WB, IH.
Immunohistochemistry:1:10

Formalin fixed (fixed <4 weeks)

Immunoblotting: 1:10

ELISA: 1:10

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Physical form

Liquid

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Okabe et al.
Cell, 159(3), 584-596 (2014-11-25)
Vascular and nervous systems, two major networks in mammalian bodies, show a high degree of anatomical parallelism and functional crosstalk. During development, neurons guide and attract blood vessels, and consequently this parallelism is established. Here, we identified a noncanonical neurovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service