Skip to Content
Merck
All Photos(1)

Documents

5.30617

Sigma-Aldrich

TRPM Antagonist, M8-An

Synonym(s):

TRPM Antagonist, M8-An, TRPM8 Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16F4N2O4
Molecular Weight:
400.32
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

10.9 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, readily bioavailable 2-picolinamide derivative that acts as a highly potent antagonist of transient receptor potential cation channel subfamily M member 8 (TRPM8; IC50 = 10.9 nM in icilin evoked Ca2+ currents in HEK293 cells stably expressing human TRPM8). At much higher levels it is shown to activate human TRPA1 (EC50 = 2.8 µM) and inhibit human TRPV1 (IC50 = 3.0 µM). Selectively reduces behavioral and neural responses to cooling in spinal ligated rats (~ 30 mg/kg), but not in naïve rats.
A cell-permeable, readily bioavailable 2-picolinamide derivative that acts as a highly potent antagonist of transient receptor potential cation channel subfamily M member 8 (TRPM8; IC50 = 10.9 nM in icilin evoked Ca2+ currents in HEK293 cells stably expressing human TRPM8). At much higher levels it is shown to activate human TRPA1 (EC50 = 2.8 µM) and inhibit human TRPV1 (IC50 = 3.0 µM). Selectively reduces behavioral and neural responses to cooling in spinal ligated rats (~ 30 mg/kg), but not in naïve rats.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
TRPM8
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Patel, R., et al. 2014. J. Pharm. Exp. Ther.345, in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service