Skip to Content
Merck
All Photos(1)

Key Documents

521276

Sigma-Aldrich

PDK1 Inhibitor II

The PDK1 Inhibitor II, also referenced under CAS 850717-64-5, controls the biological activity of PDK1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PDK1 Inhibitor II, 2-Oxo-3-[1-(1H-pyrrol-2-yl)ethylidene]indolin-5-yl]urea, BX-517

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4O2
CAS Number:
Molecular Weight:
282.30
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark brown

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H14N4O2/c1-8(11-3-2-6-17-11)13-10-7-9(18-15(16)21)4-5-12(10)19-14(13)20/h2-7,17H,1H3,(H,19,20)(H3,16,18,21)/b13-8-

InChI key

DFURSNCTQGJRRX-JYRVWZFOSA-N

General description

A cell-permeable indolinone compound that acts as a potent, ATP site-targeting PDK1 inhibitor (IC50 = 6 nM) and effectively suppresses cellular Akt Thr308 phosphorylation level in PC-3 cultures (IC50 ≤1 µM), while affecting PKA (IC50 = 1.6 µM) and 7 other kinases (IC50 ≥600 nM) only at much higher concentrations. Its in vivo applicability is limited by a low aqueous solubility (2 µg/ml in PBS) and poor pharmacokinetics.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Islam, I., et al. 2007. Bioorg. Med. Chem. Lett.17, 3819.
Islam, I., et al. 2007. Bioorg. Med. Chem. Lett.17, 3814.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service