Skip to Content
Merck
All Photos(1)

Documents

324842

Sigma-Aldrich

EGFR/ErbB-2/ErbB-4 Inhibitor II - CAS 944341-54-2 - Calbiochem

The EGFR/ErbB-2/ErbB-4 Inhibitor II, also referenced under CAS 944341-54-2, controls the biological activity of EGFR/ErbB-2/ErbB-4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

EGFR/ErbB-2/ErbB-4 Inhibitor II - CAS 944341-54-2 - Calbiochem, (E)-4-Amino-6-(4-benzyloxy-3-chloro-phenylamino)-pyrimidine-5-carbaldehyde-O-(2-morpholin-4-yl-ethyl)-oxime, HCl, JNJ-28871063, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27ClN6O3 · xHCl
CAS Number:
Molecular Weight:
482.96 (free base basis)
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

beige

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable aminopyrimidine oxime compound that potently inhibits EGFR/ErbB1/HER1, ErbB2/HER2/neu, and ErbB4/HER4 kinase activity (IC50 = 22, 38 and 21 nM, respectively) in an ATP-competitive manner and effectively suppresses cellular receptor phosphorylation (IC50 = 223 and 107 nM against basal HER2 and EGF-induced EFGR phosphorylation, respectively, in SKBR3 and A431 cultures) as well as HER2- and EGFR-dependent cancer cell proliferations (IC50<1µM), while exhibiting much reduced or little activity against a panel of 202 other kniases. Reported to be orally available and blood-brain barrier permeant in both mice and rats and be efficacious in inhibiting tumor growth in several murine xenograft models (100 to 200 mg/kg/day, p.o.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Emanuel, S.L., et al. 2008. Mol. Pharmacol.73, 338.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service