Skip to Content
Merck
All Photos(1)

Documents

200485

Millipore

Bestatin Methyl Ester - CAS 65322-89-6 - Calbiochem

Synonym(s):

Bestatin Methyl Ester - CAS 65322-89-6 - Calbiochem, (-)-N-[(2S, 3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine Methyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O4
CAS Number:
Molecular Weight:
322.40
UNSPSC Code:
12352200

Assay

≥97% (TLC)

Quality Level

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellowish

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

This product has been discontinued.



A cell-permeable derivative of Bestatin (Cat. No. 200484) that displays slightly stronger inhibition of neutral aminopeptidase than Bestatin but has much weaker activity against basic aminopeptidase. Also augments Fas- or TNF-α-induced apoptosis in solid tumor cell lines.

A cell-permeable derivative of Bestatin (Cat. No. 200484) that displays stronger inhibition of neutral aminopeptidase than Bestatin (IC50 = 31.6 nM). Exhibits weaker activity against basic aminopeptidase (IC50 = 400.1 nM). Also reported to augment Fas- (IC50 = 235.4 nM) or TNF-α-induced apoptosis in solid tumor cell lines.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Neutral amino pepsidase
Product does not compete with ATP.
Reversible: no
Target IC50: 31.6 nM against neutral amino pepsidase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Unstable in aqueous solution; dilute just prior to use.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Sekine, K., et al. 2001. Int. J. Cancer94, 485.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service