Z744015
HIGH POWER EXPANDED PLASMA CLEANER
AC/DC input 115 V, US 3-pin plug
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
PDC-001-HP (Harrick Plasma)
H × W × D
11 in. × 18 in. × 9 in.
weight
37
Looking for similar products? Visit Product Comparison Guide
General description
Our Expanded Cleaner is a larger tabletop plasma instrument with four times the capacity of the basic plasma cleaner, extensively used for nanoscale surface cleaning and surface activation.
- Larger, tabletop unit
- Adjustable RF power (Low, Medium, and High power settings)
- Applies a maximum of 30W to the RF coil
- Includes a 6" diameter x 6.5" length Pyrex chamber
- Hinged door with viewing window
- Active fan cooling
- Integral switch for a vacuum pump
- 1/8" NPT metering valve to qualitatively control gas flow and chamber pressure
- 1/8" NPT 3-way valve to quickly switch from introducing gas, venting and isolating the chamber
Other Notes
Requirements
Our Plasma Cleaners require a gas-compatible vacuum pump with minimum pump speed of 23 L /min and ultimate total pressure of
200 mTorr or less
Our Plasma Cleaners require a gas-compatible vacuum pump with minimum pump speed of 23 L /min and ultimate total pressure of
200 mTorr or less
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service