Z235768
Radioactive And Stable Isotope Tracers In Biomedicine: Principles and Practice of Kinetic Analysis
All Photos(1)
About This Item
ISBN-10:
0471561312
ISBN-13:
978-047156131-6
UNSPSC Code:
55101509
Recommended Products
publication info
R. Wolfe, John Wiley & Sons, Ltd, 1992, 480 pp., hard cover
manufacturer/tradename
Wiley
General description
A detailed, authoritative overview regarding all aspects of the theory and practice involved in utilizing isotopic tracers for the in vivo study of human metabolic function. Presents specific technical information on performing tracer studies including isotope infusion, analysis and interpretation of data.
Table of Contents
PARTIAL TABLE OF CONTENTS:
Basic Characterisitics of Isotopic Tracers.
Terminology for Tracer Studies.
Gas-Chromatography Mass-Spectrometry: Instrumentation.
Separation of Compounds by Chromatographic Techniques.
Calculation of Substrate Kinetics: Multiple-Pool Models.
Selection of Tracer Infusion and Sampling Sites.
Assessment of Body Composition.
Technical Aspects of Performing Tracer Infusions.
Glucose Metabolism.
Lipid Kinetics.
Specific Application: UREA.
Amino Acid Metabolism.
Protein Synthesis and Breakdown.
Laboratory Methods.
Basic Characterisitics of Isotopic Tracers.
Terminology for Tracer Studies.
Gas-Chromatography Mass-Spectrometry: Instrumentation.
Separation of Compounds by Chromatographic Techniques.
Calculation of Substrate Kinetics: Multiple-Pool Models.
Selection of Tracer Infusion and Sampling Sites.
Assessment of Body Composition.
Technical Aspects of Performing Tracer Infusions.
Glucose Metabolism.
Lipid Kinetics.
Specific Application: UREA.
Amino Acid Metabolism.
Protein Synthesis and Breakdown.
Laboratory Methods.
Choose from one of the most recent versions:
Certificates of Analysis (COA)