WHA110637
Whatman® polycarbonate Nuclepore AOX filter discs 0.4 μm pore size
hydrophilic, track-etched, AOX analysis, 100 ea, 25 mm diam
Synonym(s):
Whatman fitler papers, Whatman membrane, Z699993, nuclepore
About This Item
Recommended Products
Product Name
Whatman® Nuclepore™ Track-Etched Membranes, diam. 25 mm, pore size 0.4 μm, polycarbonate - AOX
material
polycarbonate - AOX
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 110637
Whatman Article No. 28417596 (US reference)
diam.
25 mm
pore size
0.4 μm
Looking for similar products? Visit Product Comparison Guide
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Features and Benefits
- High chemical resistance and thermal stability for a wide range of samples
- Low, consistent ash levels and low tare weights
- Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
- Smooth flat surface increases particle visibility
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service